missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPR Antibody (OTI4F5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-03256
This item is not returnable.
View return policy
Description
SPR Monoclonal specifically detects SPR in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| SPR | |
| Monoclonal | |
| 0.31 mg/mL | |
| Western Blot 1:500-2000, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:150 | |
| SPR | |
| Full length human recombinant protein of human SPR (NP_003115) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| 6697 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI4F5 | |
| Unconjugated | |
| EC 1.1.1.153, SDR38C1, sepiapterin reductase, sepiapterin reductase (7,8-dihydrobiopterin:NADP+ oxidoreductase), short chain dehydrogenase/reductase family 38C, member 1 | |
| Mouse | |
| 27.9 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction