missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SPR Monoclonal Antibody (OTI4F5)
GREENER_CHOICE

Product Code. 15934914
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15934914 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15934914 Supplier Invitrogen™ Supplier No. MA525308

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SPR Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

SPR encodes an aldo-keto reductase that catalyzes the NADPH-dependent reduction of pteridine derivatives and is important in the biosynthesis of tetrahydrobiopterin (BH4). Mutations in this gene result in DOPA-responsive dystonia due to sepiaterin reductase deficiency. A pseudogene has been identified on chromosome 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPR
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI4F5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SPR
Gene Accession No. P18297, P35270
Gene Alias AA409688; ENSMUSG00000071345; Gm10328; SDR38C1; sepiapterin reductase; sepiapterin reductase (7,8-dihydrobiopterin:NADP+ oxidoreductase); short chain dehydrogenase/reductase family 38C, member 1; SPR
Gene Symbols SPR
Host Species Mouse
Immunogen Full length human recombinant protein of SPR produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29270, 6697
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.