Learn More
Invitrogen™ SPR Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121157
Description
Positive test controls include: HepG2, HeLa, COS-7, Jurkat, MCF7, Mouse liver, Mouse kidney, Mouse pancreas, Rat kidney. The target is usually found in the following locations: Cytoplasm. Immunogen sequence: PLDTDMQQLA RETSVDPDMR KGLQELKAKG KLVDCKVSAQ KLLSLLEKDE FKSGAHVDFY DK.
SPR encodes an aldo-keto reductase that catalyzes the NADPH-dependent reduction of pteridine derivatives and is important in the biosynthesis of tetrahydrobiopterin (BH4). Mutations in this gene result in DOPA-responsive dystonia due to sepiaterin reductase deficiency. A pseudogene has been identified on chromosome 1.
Specifications
| SPR | |
| Polyclonal | |
| Unconjugated | |
| SPR | |
| AA409688; ENSMUSG00000071345; Gm10328; SDR38C1; sepiapterin reductase; sepiapterin reductase (7,8-dihydrobiopterin:NADP+ oxidoreductase); short chain dehydrogenase/reductase family 38C, member 1; SPR | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 20751, 29270, 6697 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.66 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P18297, P35270, Q64105 | |
| SPR | |
| A synthetic peptide corresponding to a sequence within amino acids 200-261 of human SPR (P35270). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.