Learn More
SPRED3 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41091AF594
This item is not returnable.
View return policy
Description
SPRED3 Polyclonal antibody specifically detects SPRED3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Western Blot, ELISA, Immunofluorescence |