missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPRED3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41091-0.025mg
This item is not returnable.
View return policy
Description
SPRED3 Polyclonal specifically detects SPRED3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| SPRED3 | |
| Polyclonal | |
| Unconjugated | |
| Eve-3, spred-3, Sprouty-Related, EVH1 Domain Containing 3, Sprouty-Related, EVH1 Domain-Containing Protein 3 | |
| Rabbit | |
| 45 kDa | |
| 0.025 mg | |
| Primary | |
| This Spred3 antibody is predicted to have no cross-reactivity to Spred1 or Spred2. | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.02% Sodium Azide | |
| SPRED3 | |
| Antibody was raised against a 16 amino acid synthetic peptide near the center of the human Spred3. The immunogen is located within amino acids 140 - 190 of Spred3. Amino Acid Squence: SHSRQETPPSAAAAPI | |
| Affinity Purified | |
| RUO | |
| 399473 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction