Learn More
Edmund Bühler GmbH™ Spring Clamp
Click to view available options
Holds:
10 mL Flask
500 mL
Unit Size:
Each
Description
- Stainless steel construction
- For fastening Erlenmeyer flasks on the universal tray
- The sizes are related to Erlenmeyer flasks, but are also suitable for round-bottom flasks, beakers, etc
Specifications
Specifications
| Holds | 500 mL |
| Product Type | Spring Clamp |
| Material | Stainless Steel |