Learn More
Invitrogen™ SPRR1A Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110423
This item is not returnable.
View return policy
Description
Immunogen sequence: CPSTVTPAPA QQKTKQK.
Cross-linked envelope protein of keratinocytes. It is a keratinocyte protein that first appears in the cell cytosol, but ultimately becomes cross-linked to membrane proteins by transglutaminase. All that results in the formation of an insoluble envelope beneath the plasma membrane.
Specifications
| SPRR1A | |
| Polyclonal | |
| Unconjugated | |
| SPRR1A | |
| 19 kDa pancornulin; AI528815; cornifin A; cornifin-A; mSPRR1A; small proline rich protein 1A; small proline-rich protein 1A; small proline-rich protein 1A-like; small proline-rich protein 1B (cornifin); small proline-rich protein IA; Spr; SPR1 A; Spr1a; SPR-IA; SPRK; Sprr; Sprr1; Sprr1a; Sprr1al; Sprr1b | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 20753, 6698 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.14 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P35321, Q62266 | |
| SPRR1A | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |