missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPRR2A Antibody (OTI4B3), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74347AF405
This item is not returnable.
View return policy
Description
SPRR2A Monoclonal specifically detects SPRR2A in Human samples. It is validated for Western Blot.
Specifications
| SPRR2A | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SPRR2A | |
| Full length recombinant protein of human SPRR2A (NP_005979) produced in E.coli. | |
| 0.1 mL | |
| 6700.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI4B3 | |
| Western Blot | |
| 2-1, small proline-rich protein 2A, SPR-2A | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction