missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPRY1 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99516AF532
This item is not returnable.
View return policy
Description
SPRY1 Polyclonal antibody specifically detects SPRY1 in Human samples. It is validated for Western Blot
Specifications
| SPRY1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 532 | |
| Drosophila, homolog of, 1 (antagonist of FGF signaling), sprouty homolog 1, antagonist of FGF signaling (Drosophila), spry-1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human SPRY1. | |
| 0.1 mL | |
| Cancer, Growth and Development, Neuronal Cell Markers, Signal Transduction | |
| 10252 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction