missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPRY2 Antibody (SR1722), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21915-100ul
This item is not returnable.
View return policy
Description
SPRY2 Monoclonal antibody specifically detects SPRY2 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Specifications
| SPRY2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| SR1722 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| hSPRY2, MGC23039, protein sprouty homolog 2, sprouty (Drosophila) homolog 2, sprouty 2, sprouty homolog 2 (Drosophila), spry-2 | |
| A synthesized peptide derived from human SPRY2 (Uniprot #: O43597) | |
| 100 μg | |
| Growth and Development, Neuronal Cell Markers, Signal Transduction | |
| 10253 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction