missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPRY3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119434
This item is not returnable.
View return policy
Description
SPRY3 Polyclonal Antibody for Western Blot, IHC (P)
May function as an antagonist of fibroblast growth factor (FGF) pathways and may negatively modulate respiratory organogenesis.
Specifications
| SPRY3 | |
| Polyclonal | |
| Unconjugated | |
| SPRY3 | |
| antagonist of FGF signaling; Gm1409; Gm391; Protein sprouty homolog 3; RGD1562172; sprouty homolog 3; sprouty homolog 3 (Drosophila); sprouty RTK signaling antagonist 3; sprouty3; SPRY3; spry-3 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 10251, 236576, 498159 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O43610 | |
| SPRY3 | |
| Recombinant protein Sprouty Homolog 3. The antigen corresponds to amino acid range 91-288 of the target protein. | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction