missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPRY3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5107028
This item is not returnable.
View return policy
Description
Sequence homology: Cow: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
May function as an antagonist of fibroblast growth factor (FGF) pathways and may negatively modulate respiratory organogenesis.
Specifications
| SPRY3 | |
| Polyclonal | |
| Unconjugated | |
| SPRY3 | |
| antagonist of FGF signaling; Gm1409; Gm391; Protein sprouty homolog 3; RGD1562172; sprouty homolog 3; sprouty homolog 3 (Drosophila); sprouty RTK signaling antagonist 3; sprouty3; SPRY3; spry-3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10251 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| O43610 | |
| SPRY3 | |
| Synthetic peptide directed towards the N terminal region of human SPRY3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction