missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPRY3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121579
This item is not returnable.
View return policy
Description
SPRY3 Polyclonal Antibody for Western Blot, IHC (P)
May function as an antagonist of fibroblast growth factor (FGF) pathways and may negatively modulate respiratory organogenesis.
Specifications
| SPRY3 | |
| Polyclonal | |
| Unconjugated | |
| SPRY3 | |
| antagonist of FGF signaling; Gm1409; Gm391; Protein sprouty homolog 3; RGD1562172; sprouty homolog 3; sprouty homolog 3 (Drosophila); sprouty RTK signaling antagonist 3; sprouty3; SPRY3; spry-3 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 10251, 236576, 498159 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O43610 | |
| SPRY3 | |
| Recombinant protein Sprouty Homolog 3. The antigen corresponds to amino acid range 91-288 of the target protein. | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu