missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPRYD4 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77100MFV610
This item is not returnable.
View return policy
Description
SPRYD4 Polyclonal antibody specifically detects SPRYD4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| SPRYD4 | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| DKFZp686N0877, SPRY domain containing 4, SPRY domain-containing protein 4 | |
| Antibody was raised against an 18 amino acid synthetic peptide near the center of human SPRYD4. The immunogen is located within amino acids 70 - 120 of SPRYD4. Amino Acid Squence: VTSGRHYWEVTVKRSQQF | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 283377 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction