missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SPRYD4 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA562794

Product Code. 15760582

  • € 572.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: FARSLRLCRW GAKRLGVAST EAQRGVSFKL EEKTAHSSLA LFRDDTGVKY GLVGLEPTKV ALNVERFREW AVVLADTAVT SGRHY Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 81%.

The SPRY domain-containing protein 4 (SPRYD4) is a member of a family of proteins whose sole common characteristic is the presence of a SPRY domain. SPRY domains are structural domains that were first described in the fungal Dictyostelium discoideum tyrosine kinase spore lysis A. In most systems SPRY domains provide binding sites for regulatory proteins or intramolecular binding sites that maintain the structural integrity of a protein. SPRYD4 is ubiquitously expressed and is most abundant in kidney, brain, bladder, thymus and stomach. Little is known of the function of the SPRYD4 protein.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SPRYD4
Polyclonal
Unconjugated
SPRYD4
4633402N23Rik; AA114801; RGD1306649; SPRY domain containing 4; SPRY domain-containing protein 4; Spryd4
Rabbit
Antigen affinity chromatography
RUO
283377
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8WW59
SPRYD4
Recombinant protein corresponding to Human SPRYD4. Recombinant protein control fragment (Product #RP-97853).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.