missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SPSB1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17148353
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17148353 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17148353 Supplier Invitrogen™ Supplier No. PA5110426

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MGQKVTGGIK TVDMRDPTYR PLKQELQGLD YCKPTRLDLL LDMPPVSYDV QLLHSWNNND RSLNVFVKED DKLIFHRHPV AQSTDAIRGK VGYTRGLHVW QITWAMRQRG THAVVGVATA DAPLHSVGYT TLVGNNHESW GWDLGRNRLY HDGKNQPSKT YPAFLEPDET FIVPDSFLVA LDMDDGTLSF IVDGQYMGVA FRGLKGKKLY PVVSAVWGHC EIRMRYLNGL DPEPLPLMDL CRRSVRLALG RERLGEIHTL PLPASLKAYL LYQ.

Substrate recognition component of a SCF-like ECS (Elongin BC-CUL2/5-SOCS-box protein) E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins (PubMed:15601820, PubMed:21199876). Negatively regulates nitric oxide (NO) production and limits cellular toxicity in activated macrophages by mediating the ubiquitination and proteasomal degradation of NOS2 (PubMed:21199876). Acts as a bridge which links NOS2 with the ECS E3 ubiquitin ligase complex components ELOC and CUL5 (PubMed:21199876). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPSB1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.39 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SPSB1
Gene Accession No. Q9D5L7
Gene Alias 1110014L01Rik; 4930422J18Rik; AI596360; AI854583; CBG; fi35c01; hypothetical protein LOC334190; novel SPRY domain containing protein; RGD1309319; splA/ryanodine receptor domain and SOCS box containing 1; SPRY domain-containing SOCS box protein 1; SPRY domain-containing SOCS box protein SSB-1; Spsb1; SSB1; SSB-1; Unknown (protein for IMAGE:8228399); wu:fi35c01; zgc:114115
Gene Symbols SPSB1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-273 of human SPSB1 (Q96BD6).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 74646
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.