missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SPSB2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15868903
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15868903 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15868903 Supplier Invitrogen™ Supplier No. PA570496

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 92%; Human: 100%; Mouse: 86%; Pig: 100%; Rabbit: 100%; Rat: 86%.

This gene encodes a member of a subfamily of proteins containing a central SPRY (repeats in splA and RyR) domain and a C-terminal suppressor of cytokine signaling (SOCS) box. This protein plays a role in cell signaling. This gene is present in a gene-rich cluster on chromosome 12p13 in the vicinity of the CD4 antigen and triosephosphate isomerase genes. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPSB2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SPSB2
Gene Accession No. Q99619
Gene Alias AI461677; C9; gene rich cluster, C9 gene protein; gene-rich cluster protein C9; Grcc9; splA/ryanodine receptor domain and SOCS box containing 2; SPRY domain-containing SOCS box protein 2; SPRY domain-containing SOCS box protein SSB-2; Spsb2; SSB2; SSB-2
Gene Symbols SPSB2
Host Species Rabbit
Immunogen synthetic peptide directed towards the C terminal region of human SPSB2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84727
Target Species Human, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.