missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPT3 Antibody (OTI1A5), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74348H
This item is not returnable.
View return policy
Description
SPT3 Monoclonal specifically detects SPT3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| SPT3 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| SUPT3H | |
| Full length recombinant protein of human SPT3 (NP_852001) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1A5 | |
| Western Blot, Immunohistochemistry | |
| SPT3-like protein, SPT3SPT3L, suppressor of Ty (S.cerevisiae) 3 homolog, suppressor of Ty 3 homolog (S. cerevisiae), transcription initiation protein SPT3 homolog | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication | |
| 8464.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction