Learn More
Spt6 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2584F
This item is not returnable.
View return policy
Description
Spt6 Polyclonal antibody specifically detects Spt6 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)