missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPTBN1 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67978-1-IG-150UL
This item is not returnable.
View return policy
Description
The SPTBN1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SPTBN1. This antibody recognizes human, mouse, rat, rabbit, pig antigen. The SPTBN1 antibody has been validated for the following applications: WB, IHC, ELISA analysis.Specifications
| SPTBN1 | |
| Monoclonal | |
| 1000 μg/mL | |
| ELF; SPTB2; betaSpII | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat, Rabbit, Pig | |
| 250-275 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, ELISA | |
| 1E1D1 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 6711 | |
| Store at -20°C. | |
| Monoclonal Antibody | |
| IgG1 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion