missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPTBN4 Mouse anti-Human, FITC, Clone:S393-2, Abnova™
Mouse monoclonal antibody raised against partial recombinant human SPTBN4.
Brand: Abnova MAB17113.100ug
This item is not returnable.
View return policy
Specifications
| SPTBN4 | |
| Monoclonal | |
| FITC | |
| Immunocytochemistry Immunofluorescence Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) Western Blot (1:1000) The optimal working dilution should be determined by the end user. |
|
| SPTBN4 | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| S393-2 | |
| spectrin, beta, non-erythrocytic 4 | |
| In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
| KIAA1642, QV, SPNB4, SPTBN3 | |
| Recombinant protein corresponding to amino acids 1621-1832 at C-terminus of human SPTBN4. | |
| 57731 | |
| Store at -20°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction