missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SpUlp2 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-23620AF594
This item is not returnable.
View return policy
Description
SpUlp2 Polyclonal antibody specifically detects SpUlp2 in Yeast samples. It is validated for Western Blot
Specifications
| SpUlp2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Yeast | |
| Purified |
| Western Blot | |
| Alexa Fluor 594 | |
| S. pombe Ubiquitin-like Protein 2, SpUlp -2 | |
| Reacts with residues 622-635 [SNNERQSLSSGSND] of the SpUlp2 protein. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction