Learn More
SQSTM1 Mouse anti-Human, Clone: OTI4C8, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF502133
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a multifunctional protein that binds ubiquitin and regulates activation of the nuclear factor kappa-B signaling pathway. The protein functions as a scaffolding/adaptor protein in concert with TNF receptor-associated factor 6 to mediate activation of NF-kB in response to upstream signals. Alternatively spliced transcript variants encoding either the same or different isoforms have been identified for this gene. Mutations in this gene result in sporadic and familial Paget disease of bone.Specifications
| SQSTM1 | |
| Monoclonal | |
| Unconjugated | |
| SQSTM1 | |
| A170, DMRV, FTDALS3, NADGP, OSIL, PDB3, ZIP3, p60, p62, p62B | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8878 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI4C8 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q13501 | |
| SQSTM1 | |
| Full length human recombit protein of human SQSTM1 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.