Learn More
Invitrogen™ SQSTM1 Recombinant Rabbit Monoclonal Antibody (5X4R1)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA542726
Description
Positive test controls include: 293T, HeLa, L929, C2C12, PC-12. The target is usually found in the following locations: Cytoplasm, Cytoplasmic vesicle, Endoplasmic reticulum, Late endosome, Lysosome, Nucleus, P-body, autophagosome. Immunogen sequence: LSSKEVDPST GELQSLQMPE SEGPSSLDPS QEGPTGLKEA ALYPHLPPEA DPRLIESLSQ MLSMGFSDEG GWLTRLLQTK NYDIGAALDT IQYSKHPPPL.
This gene encodes a multifunctional protein that binds ubiquitin and regulates activation of the nuclear factor kappa-B signaling pathway. The protein functions as a scaffolding/adaptor protein in concert with TNF receptor-associated factor 6 to mediate activation of NF-kB in response to upstream signals. Alternatively spliced transcript variants encoding either the same or different isoforms have been identified for this gene. Mutations in this gene result in sporadic and familial Paget disease of bone.
Specifications
| SQSTM1 | |
| Recombinant Monoclonal | |
| 1.10 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| O08623, Q13501, Q64337 | |
| Sqstm1 | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 5X4R1 | |
| Unconjugated | |
| Sqstm1 | |
| A170; EBI3-associated protein of 60 kDa; EBI3-associated protein p60; EBIAP; FTDALS3; hypothetical protein 1; ORCA; OSF-6; Osi; OSIL; oxidative stress induced; oxidative stress induced like; p60; p62; p62B; PDB3; phosphotyrosine independent ligand for the Lck SH2 domain p62; phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa; PKC-zeta-interacting protein; protein kinase C-zeta-interacting protein; sb:cb621; Sequestosome 1; sequestosome-1; Sqstm1; STAP; STONE14; ubiquitin-binding protein p62; Unknown (protein for MGC:127197); zgc:85784; ZIP; ZIP3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 113894, 18412, 8878 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.