Learn More
Kartell™ Square Bottles
Product Code. 10768335
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
Pack of 10
Pack of 100
Pack of 400
Pack of 50
Product Code. 10768335
Brand: Kartell™ 0061400
Space saving bottles, ideal for long term storage of solid or liquid samples, even with medium-high viscosity, pasty or granular materials. Easy grip caps can be wired down for security. Accurate graduation and semi-transparent walls allow a clear and precise reading of the content.
  • Seal granted by quality of both cap and mouth thread, presence of undercap.
  • Permanent graduation printed in relief externally.
  • SUITABLE FOR FOODSTUFF ACCORDING TO EC Regulation 10/2011.
  • They are safe from the point of view of the raw materials used according to 1907/2006 (REACH), BSE / TSE and 2002/95/EC (RoSH).
Specifications
Capacity (Metric) 2000 mL
Capacity (English) 67.6 oz.
Color Natural
Material Polyethylene
Sterility Non-sterile
Certifications/Compliance EC Regulation 10/2011, 1907/2006 (REACH), BSE / TSE, 2002/95/EC (RoSH)
Closure Included Yes
Disposable Reusable
Graduated Yes
Mouth Wide, Threaded
Shape Square
Fit for Purpose Certified Quality
Product Type Graduated Square Wide Neck Bottle
Autoclavable Not Autoclavable