Learn More
Huenersdorf™ Square Polyethylene Containers with Slip Lids
Product Code. p-7146945
Change view
Click to view available options
Capacity (Metric):
500 mL
1000 mL
1500 mL
2000 mL
3200 mL
Dimensions (L x W x H):
103 x 103 x 64 mm
103 x 208 x 64 mm
103 x 208 x 94 mm
208 x 208 x 64 mm
208 x 208 x 94 mm
103 x 103 x 127 mm
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Dimensions (L x W x H) unitSize
10052394 500 mL 103 x 103 x 64 mm Each
10367674 1000 mL 103 x 103 x 127 mm Each
10286174 1000 mL 103 x 208 x 64 mm Each
10516754 1500 mL 103 x 208 x 94 mm Each
10516564 2000 mL 208 x 208 x 64 mm Each
10597704 3200 mL 208 x 208 x 94 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10052394 Supplier Huenersdorf™ Supplier No. 910100
Only null left
Add to basket
Add to basket
These containers with hooded lids are made of fracture-proof, natural colored and food-safe polyethylene (HDPE and LDPE).
  • Angular shaped
  • Dustproof hooded lid, made of LDPE, reliably protects contents from contamination
  • Stackable for space-saving storage
  • Very resistant to acids, alkalis and other chemicals
  • Suitable for dishwashers, microwaves and deep freezers
  • Resistant to temperatures from −20° to +70°C
  • Printing and labeling available on request
Specifications
Capacity (Metric) 500 mL
Closure Material LDPE
Closure Type Hooded Lid
Color Natural
Material Polyethylene
Shape Square
For Use With (Application) Food Safe
For Use With (Equipment) Deep Freezer, Dishwasher, Microwave
Includes Tightly Closing LDPE Lid
Mounting Type Stackable
Product Type Container
Stackable Yes
Temperature Range (Metric) -20° to +70°C
Dimensions (L x W x H) 103 x 103 x 64 mm
Length (Metric) 103 mm
Width (Metric) 103 mm
Height (Metric) 64 mm