Learn More
Witeg™ Squibb Borosilicate Glass Separatory Funnels
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Unit Size:
Pack of 2
Description
- Acc. to Squibb
- Borosilicate glass
- Detachable PTFE stopcock
- Pear form
Specifications
Specifications
| Capacity (Metric) | 1000 mL |
| Material | Borosilicate Glass |
| Joint Number | ST 29/32 |
| Length (Metric) Stem | 100mm |
| Stopcock | Yes |
| Stopper Material | PTFE |
| Stopper Number | ST 29/32 |
| Product Type | Separatory Funnel |