missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SR 16832

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
SR 16832 is a dual site PPARγ inhibitor. Acts at orthosteric and allosteric sites in the ligand binding domain. Inhibits binding of endogenous ligands and transcriptional activity of PPARγ, more effectively than the orthosteric covalent antagonists GW 9662 (Cat. No. 1508) and T 0070907 (Cat. No. 2301).
Specifications
Specifications
| CAS | 2088135-12-8 |
| Synonym | 2-Chloro-N-(6-methoxy-4-quinolinyl)-5-nitrobenzamide |
| Quantity | 5 mg |
| Molecular Formula | C17H12ClN3O4 |
| Formula Weight | 357.75 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur