missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SR-AI/MSR Antibody (268318), CoraFluor™ 1, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB1797CL1
This item is not returnable.
View return policy
Description
SR-AI/MSR Monoclonal antibody specifically detects SR-AI/MSR in Mouse samples. It is validated for Flow Cytometry,Western Blot,CyTOF
Specifications
| SR-AI/MSR | |
| Monoclonal | |
| CoraFluor 1 | |
| CD204, CD204 antigen, Macrophage acetylated LDL receptor I and II, macrophage scavenger receptor 1, macrophage scavenger receptor type III, phSR1, phSR2, SCARA1macrophage scavenger receptor types I and II, Scavenger receptor class A member 1, scavenger receptor class A, member 1, SR-A | |
| Mouse myeloma cell line NS0-derived recombinant mouse SR-AI/MSR, Trp79-Ser454, Accession # AAA39747 | |
| 0.1 mL | |
| Adaptive Immunity, Cancer, Immunology, Innate Immunity, Lipid and Metabolism, Neuroscience, Prostate Cancer | |
| 4481 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Flow Cytometry, Western Blot, CyTOF | |
| 268318 | |
| PBS | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction