missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SR-AI/MSR Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB400-159F
This item is not returnable.
View return policy
Description
SR-AI/MSR Polyclonal specifically detects SR-AI/MSR in Human, Primate samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| SR-AI/MSR | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| P21757 | |
| MSR1 | |
| A synthetic peptide made to an internal portion of human Macrophage Scavenger Receptor I (between residues 300-350). [UniProt P21757] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human and monkey. There have been mixed results (positive and negative) with mouse. | |
| Store at 4C in the dark. |
| Immunocytochemistry, Immunofluorescence | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| CD204, CD204 antigen, Macrophage acetylated LDL receptor I and II, macrophage scavenger receptor 1, macrophage scavenger receptor type III, phSR1, phSR2, SCARA1macrophage scavenger receptor types I and II, Scavenger receptor class A member 1, scavenger receptor class A, member 1, SR-A | |
| Rabbit | |
| 50 kDa | |
| 0.1 mL | |
| Adaptive Immunity, Cancer, Immunology, Innate Immunity, Lipid and Metabolism, Neuroscience, Prostate Cancer | |
| 4481 | |
| Human, Primate | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction