missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SR-BI Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB400-104G
This item is not returnable.
View return policy
Description
SR-BI Polyclonal specifically detects SR-BI in Human, Mouse, Rat, Porcine, Chinese Hamster, Hamster, Mustelid, Primate, Rabbit, Golden Syrian Hamster samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated, Knockdown Validated.
Specifications
| SR-BI | |
| Polyclonal | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated, Knockdown Validated | |
| CD36 and LIMPII analogous 1, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 1, CD36L1scavenger receptor class B type 1, CLA1 CD36 antigen-like 1, CLA-1 HDLQTL6, Collagen type I receptor, thrombospondin receptor-like 1, MGC138242, SCARB1, scavenger receptor class B type III, scavenger receptor class B, member 1, SR-B1, SRB1 scavenger receptor class B member 1, SRBI | |
| Rabbit | |
| 82 kDa | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cardiovascular Biology, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction, Virology Bacteria and Parasites | |
| 949 | |
| Human, Mouse, Rat, Chinese Hamster, Hamster, Mustelid, Primate, Rabbit, Golden Syrian Hamster | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay, KnockDown | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SCARB1 | |
| A C-terminal peptide containing residues from mouse SR-BI (within residues 450-509). [UniProt# Q61009] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Mouse, rat, mink, hamster, primate, rabbit and the long form of human SR-BI. Reactivity with CHO cells reported in scientific literature (PMID: 12356718). | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction