missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SR-BI Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99690MFV610
This item is not returnable.
View return policy
Description
SR-BI Polyclonal antibody specifically detects SR-BI in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| SR-BI | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation | |
| CD36 and LIMPII analogous 1, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 1, CD36L1scavenger receptor class B type 1, CLA1 CD36 antigen-like 1, CLA-1 HDLQTL6, Collagen type I receptor, thrombospondin receptor-like 1, MGC138242, SCARB1, scavenger receptor class B type III, scavenger receptor class B, member 1, SR-B1, SRB1 scavenger receptor class B member 1, SRBI | |
| Produced in rabbits immunized with purified, recombinant Human SR-BI (Accession#: NP_005496.4; Pro33-Tyr443) | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cardiovascular Biology, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction, Virology Bacteria and Parasites | |
| 949 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunocytochemistry, Immunoprecipitation | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction