missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SR-BI Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5143116
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize both reported isoforms (NP_005496.4; NP_001076428.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
High density lipoproteins (HDLs) play a critical role in cholesterol metabolism and their plasma concentrations are inversely correlated with risk for atherosclerosis. The SR-BI binds HDLs and mediates selective uptake of HDL cholesteryl ester. SR-BI binds HDL with high affinity, is expressed primarily in liver and nonplacental steroidgenic tissues, and mediates selective cholesterol uptake by a distinct mechanism. In mice, it seems that SR-BI plays a key role in determining the levels of plasma lipoprotein cholesterol and the accumulation of cholesterol stores in the adrenal gland.
Specifications
| SR-BI | |
| Polyclonal | |
| Unconjugated | |
| SCARB1 | |
| AI120173; CD36; CD36 and LIMPII analogous 1; CD36 antigen (collagen type I receptor thrombospondin receptor)-like 1 (scavanger receptor class B type 1); CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 1; CD36 antigen (collagen type I receptor, thrombospondin receptor-like 1); CD36 antigen-like 1; Cd36l1; CD36-related class B scavenger receptor haSR-BI; CLA1; Cla-1; Collagen type I receptor, thrombospondin receptor-like 1; D5Ertd460e; haSR-bI; HDL QTL 1; Hdlq1; HDLQTL6; high density lipoprotein receptor; Hlb398; mSR-BI; Scarb1; scavanger receptor class B type 1; scavenger receptor b1; scavenger receptor class B member 1; scavenger receptor class B type I; scavenger receptor class B type II; scavenger receptor class B type III; scavenger receptor class B, member 1; scavenger receptor class B1; SRB1; SR-B1; SRBI; SR-BI; SR-BII | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 949 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q8WTV0 | |
| SCARB1 | |
| Peptide with sequence C-NGLSKVDFWHSDQ (aa 238-250). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction