Learn More
Invitrogen™ SR-BI Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA590750
Product Code. 16557981
  • € 520.00 / 100µL
Description
Immunogen sequence: MGCSAKARWA AGALGVAGLL CAVLGAVMIV MVPSLIKQQV LKNVRIDPSS LSFNMWKEIP IPFYLSVYFF DVMNPSEILK GEKPQVRERG PYVYREFRHK; Positive Samples: Rat ovary; Cellular Location: Cell membrane, Membrane, Multi-pass membrane protein, caveola.
High density lipoproteins (HDLs) play a critical role in cholesterol metabolism and their plasma concentrations are inversely correlated with risk for atherosclerosis. The SR-BI binds HDLs and mediates selective uptake of HDL cholesteryl ester. SR-BI binds HDL with high affinity, is expressed primarily in liver and nonplacental steroidgenic tissues, and mediates selective cholesterol uptake by a distinct mechanism. In mice, it seems that SR-BI plays a key role in determining the levels of plasma lipoprotein cholesterol and the accumulation of cholesterol stores in the adrenal gland.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SR-BI
Polyclonal
Unconjugated
SCARB1
AI120173; CD36; CD36 and LIMPII analogous 1; CD36 antigen (collagen type I receptor thrombospondin receptor)-like 1 (scavanger receptor class B type 1); CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 1; CD36 antigen (collagen type I receptor, thrombospondin receptor-like 1); CD36 antigen-like 1; Cd36l1; CD36-related class B scavenger receptor haSR-BI; CLA1; Cla-1; Collagen type I receptor, thrombospondin receptor-like 1; D5Ertd460e; haSR-bI; HDL QTL 1; Hdlq1; HDLQTL6; high density lipoprotein receptor; Hlb398; mSR-BI; Scarb1; scavanger receptor class B type 1; scavenger receptor b1; scavenger receptor class B member 1; scavenger receptor class B type I; scavenger receptor class B type II; scavenger receptor class B type III; scavenger receptor class B, member 1; scavenger receptor class B1; SRB1; SR-B1; SRBI; SR-BI; SR-BII
Rabbit
Affinity Chromatography
RUO
20778, 25073, 949
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
0.28 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P97943, Q61009, Q8WTV0
SCARB1
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human SR-BI (NP_0054964).