missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SR Recombinant Rabbit Monoclonal Antibody (JE61-82)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544854
This item is not returnable.
View return policy
Description
SR Recombinant Monoclonal Antibody for Western Blot, IHC (P)
SR proteins are a highly conserved family of arginine/serine-rich, spliceosome-associated phosphoproteins essential for metazoan pre-mRNA splicing. SR proteins act early in splicing by promoting splice site recognition and spliceosome assembly. SR proteins also play a regulatory role because they can determine alternative splice site usage in vivo and in vitro. SR proteins appear to be recruited from nuclear speckles, in which they are concentrated to sites of transcription in order to spatially coordinate transcription and pre-mRNA splicing within the cell nucleus.
Specifications
| SR | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| G3V6S8, O35326, P84103, P84104, Q08170, Q09167, Q13243, Q13247, Q3TWW8, Q8VE97 | |
| SRSF3, SRSF4, SRSF5, SRSF6 | |
| Synthetic peptide within human SR protein repeat aa 150-199/344. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| JE61-82 | |
| Unconjugated | |
| SRSF6 | |
| 1210001E11Rik; 5730499P16Rik; AI314910; AL024116; AW146126; AW550192; B52; Cl-4; Delayed-early protein HRS; epididymis secretory protein Li 91; HEL-S-91; HRS; hypothetical protein; insulin-induced growth response protein CL-4; LOW QUALITY PROTEIN: serine/arginine-rich splicing factor 4; MNCb-2616; pre-mRNA splicing factor SRp20; pre-mRNA-splicing factor SRP20; Pre-mRNA-splicing factor SRP40; pre-mRNA-splicing factor SRP55; Pre-mRNA-splicing factor SRP75; Protein X16; RGD1561347; serine and arginine rich splicing factor 3; serine and arginine rich splicing factor 4; serine and arginine rich splicing factor 5; serine and arginine rich splicing factor 6; serine/arginine-rich splicing factor 3; Serine/arginine-rich splicing factor 4; serine/arginine-rich splicing factor 5; Serine/arginine-rich splicing factor 6; SFRS3; Sfrs4; Sfrs5; Sfrs6; splicing factor; splicing factor arginine/serine-rich 5 (SRp40 HRS); splicing factor SRp55; splicing factor SRp55 homolog; splicing factor SRp55-1; splicing factor, arginine/serine-rich 3; splicing factor, arginine/serine-rich 3 (SRp20); splicing factor, arginine/serine-rich 4; splicing factor, arginine/serine-rich 4 (SRp75); splicing factor, arginine/serine-rich 5; splicing factor, arginine/serine-rich 5 (SRp40, HRS); splicing factor, arginine/serine-rich 6; splicing factor, arginine/serine-rich, 20-kD; splicing factor, arginine/serine-rich, 55 kDa; SR splicing factor 4; SR splicing factor 5; SR splicing factor 6; SRP001LB; Srp20; SRP40; SRP55; SRP55-1; SRP75; SRSF3; SRSF4; SRSF5; Srsf6; X16 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 20383, 20384, 29667, 361814, 362264, 362612, 57317, 6428, 6429, 6430, 6431, 67996 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction