Learn More
SR17018, MedChemExpress
Product Code. 16464458
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16464458
Brand: Medchem Express HY111454
MedChemExpress SR17018 is an mu-opioid-receptor (MOR) agonist, binding with GTPγS, with an EC50 of 97 nM.
SR17018 is an mu-opioid-receptor (MOR) agonist, binding with GTPγS, with an EC50 of 97 nM.
Specifications
Chemical Name or Material SR17018
CAS 2134602-45-0
Color White
Purity Grade Notes Research
Molecular Formula C19H18Cl3N3O
Quantity 10 mg
Solubility Information DMSO : 12.5 mg/mL (30.43 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C1NC2=CC(Cl)=C(Cl)C=C2N1C3CCN(CC4=CC=C(Cl)C=C4)CC3
Molecular Weight (g/mol) 410.72
Formula Weight 410.72
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less
Research purposes only