missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRA1 Antibody (1D4H8), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
€ 203.00
Specifications
| Antigen | SRA1 |
|---|---|
| Clone | 1D4H8 |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
SRA1 Monoclonal specifically detects SRA1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| SRA1 | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| MGC87674, pp7684, SRA, SRAP, steriod receptor RNA activator 1, steriod receptor RNA activator protein, steroid receptor coactivator, steroid receptor RNA activator 1, steroid receptor RNA activator 1 (complexes with NCOA1), Steroid receptor RNA activator protein, STRAA1 | |
| SRA1 | |
| IgG1 | |
| Unpurified |
| 1D4H8 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| 10011 | |
| Purified recombinant fragment of SRA expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title