missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SRA1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA562145

Product Code. 15769941

  • € 566.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: RMALLVQELS SHRWDAADDI HRSLMVDHVT EVSQWMVGVK RLIAEKRSLF SEEAANEEKS AATAEKNHTI PGFQQAS Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 77%.

This gene is involved in transcriptional coactivation by steroid receptors. There is currently data suggesting this gene encodes both a non-coding RNA that functions as part of a ribonucleoprotein complex and a protein coding mRNA. Increased expression of both the transcript and the protein is associated with cancer.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SRA1
Polyclonal
Unconjugated
SRA1
AA959952; LOW QUALITY PROTEIN: steroid receptor RNA activator 1; MGC87674; pp7684; RGD:621148}; Sra; sra1; sra1 {ECO:0000312; SRAP; steroid receptor coactivator; steroid receptor RNA activator 1; steroid receptor RNA activator 1 (complexes with NCOA1); steroid receptor RNA activator protein; Straa1; Strra1; Unknown (protein for MGC:128615)
Rabbit
Antigen affinity chromatography
RUO
10011
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9HD15
SRA1
Recombinant protein corresponding to Human SRA1. Recombinant protein control fragment (Product #RP-96989).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.