missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SRBD1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17168353
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17168353 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17168353 Supplier Invitrogen™ Supplier No. PA5110428

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: LGLEGAARAL LEKPGELSLL SYIRPDVKGL STLQDIEIGV QHILADMIAK DKDTLDFIRN LCQKRHVCIQ SSLAKVSSKK VNEKDVDKFL LYQHFSCNIR NIHHHQILAI NRGENLKVLT VKVNISDGVK DEFCRWCIQN RWRPRSFARP ELMKILYNSL NDSFKRLIYP LLCREFRAKL TSDAEKESVM MFGRNLRQLL LTSPVPGRTL MGVDPGYKHG CKLAIISPTS QILHTDVVYL HCGQGFREAE KIKTLLLNFN CSTVVIGNGT A.

SRBD1 gene ontology annotations related to this gene include nucleobase-containing compound metabolic process; translation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRBD1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SRBD1
Gene Accession No. Q497V5, Q8N5C6
Gene Alias AI461933; C85414; D530025C17Rik; H_NH0576F01.1; RGD1562788; S1 RNA binding domain 1; S1 RNA-binding domain-containing protein 1; Srbd1; WUGSC:H_NH0576F01.1
Gene Symbols SRBD1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 330-600 of human SRBD1 (NP_060549.4).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 301665, 55133, 78586
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.