Learn More
Src Family Kinases Antibody Pack, Novus Biologicals™
Antibody Pack
Brand:  Novus Biologicals NBP3-20215
Product Code. 18699723
  • € 947.00 / Pack of 11
Description
Src Family Kinases antibody specifically detects Src Family Kinases in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Src Family Kinases
Unconjugated
11 Vials
Human
Purified
Western Blot
See individual datasheets.
RUO
Storage is content dependent.