missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Src, p Tyr419 Antibody (S01-6H3), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19758-100ul
This item is not returnable.
View return policy
Description
Src Monoclonal antibody specifically detects Src in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Src | |
| Monoclonal | |
| 0.3 mg/mL | |
| Western Blot 1:1000 | |
| c-src, EC 2.7.10, EC 2.7.10.2, pp60c-src, Rous sarcoma, tyrosine kinase pp60c-src, tyrosine-protein kinase SRC-1, v-src avian sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog, v-src sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog (avian) | |
| A synthetic phosphopeptide corresponding to residues surrounding Tyr419 of human Src (Uniprot # P12931) | |
| 100 μg | |
| Cancer Stem Cells, Phospho Specific, Protein Kinase, Signal Transduction, Transcription Factors and Regulators, Tyrosine Kinases | |
| 6714 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| S01-6H3 | |
| Unconjugated | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction