Learn More
Invitrogen™ SRD5A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120297
Description
Positive test controls include: Mouse brain, Mouse liver, Mouse testis, Rat liver. The target is usually found in the following locations: Endoplasmic reticulum membrane, Microsome membrane, Multi-pass membrane protein. Immunogen sequence: LIYPFLMRGG KPMPLLACTM AIMFCTCNGY LQSRYLSHCA VYADDWVTDP RFLIGFGLWL TGMLINIHSD HILRNLRKPG DTGYKIPRGG LFEYVTAANY F.
Steroid 5alpha-Reductase is an important enzyme in androgen physiology because it catalyzes the conversion of testosterone into the more potent 5alpha-dihydro-testosterone, which mediates androgen effects on target tissues. The enzyme exists as two isoforms: type 1, which is expressed mainly in the skin; and type 2, which is expressed mainly in the prostate. In cultured human skin cells, 5alpha-Reductase 1 shows heterogeneity of protein, and has different levels of transcriptional and translational expression. 5alpha-Reductase 1 is expressed in all portions of the hair follicle, whereas 5alpha-Reductase 2 is expressed only in mesenchymal portions. In addition, 5alpha-Reductase 1 is mainly expressed in human breast carcinoma and may play a role in the in situ production and actions of the potent androgen 5alpha-dihydrotestosterone, including inhibition of cancer cell proliferation in hormone-dependent human breast carcinoma. The 5alpha-Reductase-3alpha-hydroxysteroid dehydrogenase complex is present in the human brain, suggesting that the complex may be involved in the synthesis of neuroactive steroids or the catabolism of neurotoxic steroids.
Specifications
| SRD5A1 | |
| Polyclonal | |
| Unconjugated | |
| SRD5A1 | |
| 0610031P22Rik; 3-oxo-5-alpha-steroid 4-dehydrogenase 1; 4930435F02Rik; S5AR 1; SR type 1; Srd5a1; Srd5a-1; steroid 5 alpha-reductase 1; steroid 5-alpha-reductase 1; steroid 5-alpha-reductase type I; steroid-5-alpha-reductase 1; steroid-5-alpha-reductase, alpha polypeptide 1 (3-oxo-5 alpha-steroid delta 4-dehydrogenase alpha 1); type 1 5alpha-reductase | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 24950, 78925 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.57 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P24008, Q68FF9 | |
| SRD5A1 | |
| Synthetic peptide | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.