missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SRD5A2 Recombinant Rabbit Monoclonal Antibody (24GB2565)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557268
This item is not returnable.
View return policy
Description
SRD5A2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes a microsomal protein expressed at high levels in androgen-sensitive tissues such as the prostate. The encoded protein is active at acidic pH and is sensitive to the 4-azasteroid inhibitor finasteride. Deficiencies in this gene can result in male pseudohermaphroditism, specifically pseudovaginal perineoscrotal hypospadias (PPSH).
Specifications
| SRD5A2 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| SRD5A2 | |
| 3-oxo-5-alpha-steroid 4-dehydrogenase 2; 5 alpha-SR2; 5ART2; MGC138457; S5AR 2; SR type 2; SRD5A2; ST5AR2; steroid 5 alpha reductase type2; steroid 5 alpha-reductase 2; steroid 5-alpha-reductase 2; steroid-5-alpha-reductase, alpha polypeptide 2 (3-oxo-5 alpha-steroid delta 4-dehydrogenase alpha 2); type 2; 4-ene-3-oxosteroid 5-alpha oxidoreductase; type II 5-alpha reductase | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 64677, 6716, 94224 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB2565 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P31213, P31214, Q99N99 | |
| SRD5A2 | |
| A synthesized peptide derived from human SRD5A2 (aa 1-70). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction