missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SREBP2 Antibody (1D2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 15 publications
Brand: Novus Biologicals NBP1-54446
This item is not returnable.
View return policy
Description
SREBP2 Monoclonal specifically detects SREBP2 in Human, Chicken samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Gel Super Shift Assays.
Specifications
| SREBP2 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 2 - 5 ug/mL, ELISA, Immunocytochemistry/Immunofluorescence 10 ug/mL, Immunoprecipitation 10 ug/mg protein, Gel Super Shift Assays | |
| BHLHD2, bHLHd2sterol regulatory element-binding protein 2, Class D basic helix-loop-helix protein 2, SREBP2, SREBP2SREBP-2, sterol regulatory element binding transcription factor 2, Sterol regulatory element-binding transcription factor 2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 6721 | |
| Human, Chicken | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Gel Shift | |
| 1D2 | |
| Unconjugated | |
| Q12772 | |
| SREBF2 | |
| Human SREBP2 protein (amino acids 48-403). [UniProt# Q12772] | |
| 0.1 mL | |
| Primary | |
| Recognizes the precursor and mature forms of human SREBP2. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction