missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SREBP2 Antibody (SREBP2/1579), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08527C
This item is not returnable.
View return policy
Description
SREBP2 Monoclonal antibody specifically detects SREBP2 in Human samples. It is validated for ELISA
Specifications
| SREBP2 | |
| Monoclonal | |
| DyLight 650 | |
| BHLHD2, bHLHd2sterol regulatory element-binding protein 2, Class D basic helix-loop-helix protein 2, SREBP2, SREBP2SREBP-2, sterol regulatory element binding transcription factor 2, Sterol regulatory element-binding transcription factor 2 | |
| Recombinant full-length human SREBP2 protein (Uniprot: Q12772) | |
| 100 μg | |
| metabolism | |
| 6721 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| ELISA | |
| SREBP2/1579 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction