missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SREC-I/SCARF1 Antibody (373606) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2409JF585
This item is not returnable.
View return policy
Description
SREC-I/SCARF1 Monoclonal antibody specifically detects SREC-I/SCARF1 in Human samples. It is validated for Flow Cytometry,Western Blot,CyTOF
Specifications
| SREC-I/SCARF1 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| Acetyl LDL receptor, KIAA0149endothelial cells scavenger receptor, MGC47738, scavenger receptor class F, member 1, Scavenger receptor expressed by endothelial cells 1, SREC1, SREC-I, SRECscavenger receptor expressed by endothelial cells | |
| Mouse myeloma cell line NS0-derived recombinant human SREC-I/SCARF1, Ser20-Thr421, Accession # Q14162 | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 8578 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, Western Blot, CyTOF | |
| 373606 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction