missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRF Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16821-1-AP-150UL
This item is not returnable.
View return policy
Description
Serum response factor (SRF) is a transcription factor that binds the serum response element (SRE), a sequence that mediates the transient response of many cellular genes to growth stimulation.Specifications
| SRF | |
| Polyclonal | |
| Unconjugated | |
| SRF | |
| MCM1, Serum response factor, SRF | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20807, 501099, 6722 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P11831, Q9JM73 | |
| Srf | |
| SRF Fusion Protein Ag10386 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction