Learn More
SRF Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16821-1-AP-20UL
 View more versions of this product
Product Code. 16836574
  • € 145.00 / 20µL
Description
Serum response factor (SRF) is a transcription factor that binds the serum response element (SRE), a sequence that mediates the transient response of many cellular genes to growth stimulation.
Specifications
Specifications
SRF
Polyclonal
Unconjugated
SRF
MCM1, Serum response factor, SRF
Rabbit
Antigen Affinity Chromatography
RUO
20807, 501099, 6722
-20°C
Liquid
Western Blot, Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin)
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P11831, Q9JM73
Srf
SRF Fusion Protein Ag10386
20 μL
Primary
Human, Mouse, Rat