missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRFBP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00297-100ul
This item is not returnable.
View return policy
Description
SRFBP1 Polyclonal antibody specifically detects SRFBP1 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| SRFBP1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1-4 uL/mg of lysate | |
| BUD22, BUD22 homolog, FLJ25286, p49, p49/STRAP, Rlb1, serum response factor binding protein 1, serum response factor-binding protein 1, SRF-dependent transcription regulation associated protein, SRF-dependent transcription regulation-associated protein, STRAP | |
| Produced in rabbits immunized with purified, recombinant Human SRFBP1 (Uniprot#: Q8NEF9; Met1-Asp429) | |
| 100 μg | |
| Cardiovascular Biology | |
| 153443 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction