missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SRGAP1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15700402 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15700402 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15700402 Supplier Invitrogen™ Supplier No. PA562604

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: SPTDRHPDGY LARQRKRGEP PPPVRRPGRT SDGHCPLHPP HALSNSSVDL GSPSLASHPR GLLQNRGLNN DSPERRRRPG HGSLTNISRH DSLKKIDSPP IRRSTSSGQY TGFND Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 94%.

The protein encoded by this gene is a GTPase activator, working with the GTPase CDC42 to negatively regulate neuronal migration. The encoded protein interacts with the transmembrane receptor ROBO1 to inactivate CDC42.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRGAP1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SRGAP1
Gene Accession No. Q7Z6B7
Gene Alias 4930572H05Rik; Arhgap13; KIAA1304; NMTC2; RGD1566260; Rho GTPase activating protein 13; rho GTPase-activating protein 13; SLIT-ROBO Rho GTPase activating protein 1; SLIT-ROBO Rho GTPase-activating protein 1; Srgap1
Gene Symbols SRGAP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SRGAP1. Recombinant protein control fragment (Product #RP-101995).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57522
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.